Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 4 | 648 | ||
NM_002429.5 | NP_002420.1 | 4 | 648 | |||
NR_073606.1 | 4 | 619 | ||||
XM_011538359.1 | XP_011536661.1 | 4 | 634 | |||
XM_017019308.1 | XP_016874797.1 | 4 | 634 | |||
XM_017019309.1 | XP_016874798.1 | 4 | 634 | |||
GenBank mRNA | AK225939.1 | - | 4 | 638 | ||
AK297999.1 | - | 3 | 474 | |||
AK303202.1 | - | 4 | 601 | |||
AL545199.3 | - | 4 | 648 | |||
BC050368.1 | - | 4 | 587 | 5761074 | ||
U37791.1 | - | 4 | 576 | |||
U38321.1 | - | 4 | 571 | |||
U38431.1 | - | 4 | 540 | |||
X92521.1 | - | 4 | 596 | |||
Y08622.1 | - | 4 | 595 |