Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 3 | 527 | ||
GenBank mRNA | AK030908.1 | - | 1 | 1360 | ||
AK075579.1 | - | 3 | 556 | |||
AK154071.1 | - | 3 | 515 | |||
AK185969.1 | - | 1 | 18 | |||
BC010777.1 | - | 3 | 527 | 4241357 |