Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 7 | 900 | ||
GenBank mRNA | AK017523.1 | - | 8 | 1336 | ||
AK030908.1 | - | 2 | 2085 | |||
AK045166.1 | - | 1 | 100 | |||
AK075579.1 | - | 5 | 1200 | |||
AK154071.1 | - | 7 | 888 | |||
BC010777.1 | - | 7 | 900 | 4241357 |