Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024239.2 | NP_077201.1 | 2 | 291 | ||
GenBank mRNA | AB010123.1 | - | 2 | 238 | ||
AK017600.1 | - | 2 | 246 | |||
AK019907.1 | - | 2 | 241 | |||
AK046995.1 | - | 2 | 243 | |||
AK136961.1 | - | 2 | 842 | |||
AK139391.1 | - | 2 | 266 | |||
AK168810.1 | - | 2 | 264 | |||
AK220541.1 | - | 2 | 214 | |||
BC003497.1 | - | 2 | 234 | 3153829 | ||
BC006939.1 | - | 2 | 289 | 2649718 | ||
BC025111.1 | - | 2 | 233 | 4194332 | ||
CJ074803.1 | - | 2 | 291 |