Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027453.2 | NP_081729.1 | 4, 5 | 462 | ||
GenBank mRNA | AK004239.1 | - | 4, 5 | 493 | ||
AK011367.1 | - | 4, 5 | 808 | |||
AK013120.1 | - | 4, 5 | 450 | |||
AK017614.1 | - | 4, 5 | 423 | |||
AK076283.1 | - | 4, 5 | 462 | |||
BC024612.1 | - | 4, 5 | 445 | |||
BC024920.1 | - | 4, 5 | 405 | |||
BC058282.1 | - | 4, 5 | 418 | 30077136 | ||
BC099407.1 | - | 3, 4 | 397 | 30938785 | ||
BC138215.1 | - | 4, 5 | 443 | 8861234 | ||
BC138216.1 | - | 4, 5 | 443 | 8861235 |