Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027455.2 | NP_081731.1 | 7 | 1399 | ||
GenBank mRNA | AK017598.1 | - | 7 | 1062 | ||
AK037197.1 | - | 7 | 1052 | |||
AK045974.1 | - | 7 | 1400 | |||
AK148394.1 | - | 7 | 1070 | |||
BC020023.1 | - | 6 | 970 | 3491756 | ||
BC151027.1 | - | 7 | 1143 | 9087940 | ||
BC151029.1 | - | 7 | 1143 | 9087942 |