Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 9 | 1029 | ||
GenBank mRNA | AK017582.1 | - | 9 | 924 | ||
AK034025.1 | - | 9 | 1018 | |||
AK039945.1 | - | 9 | 896 | |||
AK046466.1 | - | 9 | 1030 | |||
AK173153.1 | - | 1526 | ||||
BC052488.1 | - | 9 | 905 | 6438726 |