Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023579.4 | NP_076068.1 | 8 | 956 | ||
GenBank mRNA | AF294327.1 | - | 8 | 907 | ||
AK017701.1 | - | 7 | 1004 | |||
AK049422.1 | - | 8 | 928 | |||
AK053681.1 | - | 8 | 921 | |||
AK079134.1 | - | 8 | 926 | |||
AK167303.1 | - | 8 | 918 | |||
AK169664.1 | - | 8 | 924 | |||
BC020012.1 | - | 2 | 228 | |||
BC052392.1 | - | 8 | 899 | 5719784 | ||
BC054814.1 | - | 8 | 1129 | 6842146 |