Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023579.4 | NP_076068.1 | 22 | 2583 | ||
GenBank mRNA | AF294327.1 | - | 22 | 2534 | ||
AK049422.1 | - | 22 | 2555 | |||
AK053681.1 | - | 22 | 2548 | |||
AK079134.1 | - | 22 | 2553 | |||
AK161668.1 | - | 16 | 1804 | |||
AK167303.1 | - | 22 | 2543 | |||
AK169664.1 | - | 22 | 2551 | |||
AK210136.1 | - | 2 | 93 | |||
BC020012.1 | - | 16 | 1855 | |||
BC051433.1 | - | 15 | 1628 | |||
BC052392.1 | - | 22 | 2526 | 5719784 | ||
BC054814.1 | - | 22 | 2756 | 6842146 | ||
BC066204.1 | - | 5 | 420 |