Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276767.1 | NP_001263696.1 | 13 | 1754 | ||
NM_144530.6 | NP_653113.4 | 12 | 1700 | |||
GenBank mRNA | AK078105.1 | - | 12 | 1663 | ||
AK122341.2 | - | 11 | 5235 | |||
AK134057.1 | - | 9 | 1180 | |||
AK145740.1 | - | 12 | 1673 | |||
AK167700.1 | - | 12 | 1674 | |||
BC066163.1 | - | 13 | 1712 | 30092453 | ||
BC066848.1 | - | 11 | 2511 | 30471872 | ||
FR682665.1 | - | 13 | 1673 |
Gene Symbol: Zbed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FR682664.1 | - | 7512 |