Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033155.1 | NP_001028327.1 | 3 | 656 | ||
GenBank mRNA | AK017735.1 | - | 3 | 518 | ||
AK139489.1 | - | 3 | 382 | |||
AK160961.1 | - | 3 | 445 | |||
BC117717.1 | - | 3 | 457 | 40088109 | ||
BC117718.1 | - | 3 | 457 | 40088110 |