Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159351.1 | NP_001152823.1 | 3 | 253 | ||
NM_023585.4 | NP_076074.2 | 4 | 379 | |||
GenBank mRNA | AF303828.1 | - | 3 | 336 | ||
AK017786.1 | - | 3 | 234 | |||
AK019486.1 | - | 4 | 350 | |||
AK028472.1 | - | 4 | 353 | |||
AK028742.1 | - | 4 | 363 | |||
AK031233.1 | - | 4 | 348 | |||
AK033016.1 | - | 4 | 364 | |||
AK146073.1 | - | 4 | 351 | |||
BC029742.1 | - | 4 | 347 | |||
BC044722.1 | - | 3 | 217 | |||
BC058374.1 | - | 4 | 350 | 5715168 | ||
BC083098.1 | - | 4 | 364 | 30460946 | ||
BC094903.1 | - | 4 | 342 | 30081846 | ||
CJ049747.1 | - | 3 | 237 |