Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159351.1 | NP_001152823.1 | 2 | 163 | ||
NM_023585.4 | NP_076074.2 | 2 | 163 | |||
GenBank mRNA | AF303828.1 | - | 1 | 120 | ||
AK017786.1 | - | 2 | 144 | |||
AK028472.1 | - | 2 | 137 | |||
AK028742.1 | - | 2 | 147 | |||
AK031233.1 | - | 2 | 132 | |||
AK033016.1 | - | 2 | 148 | |||
AK146073.1 | - | 2 | 135 | |||
BC029742.1 | - | 2 | 131 | |||
BC044722.1 | - | 2 | 127 | |||
BC058374.1 | - | 2 | 134 | 5715168 | ||
BC083098.1 | - | 2 | 148 | 30460946 | ||
BC094903.1 | - | 2 | 126 | 30081846 | ||
BY025619.1 | - | 2 | 170 | |||
CJ049747.1 | - | 2 | 147 |