Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042653.1 | NP_001036118.1 | 3 | 522 | ||
GenBank mRNA | AK017825.1 | - | 3 | 487 | ||
AK045323.1 | - | 2 | 725 | |||
BC115883.1 | - | 3 | 478 | 40088361 | ||
BC116660.1 | - | 3 | 479 | 40088363 |