Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081430.1 | NP_001074899.1 | 5 | 1201 | ||
GenBank mRNA | AK015640.1 | - | 5 | 1174 | ||
AK039333.1 | - | 2 | 2325 | |||
AK044235.1 | - | 4 | 821 | |||
BC054060.1 | - | 3 | 428 | |||
BC157931.1 | - | 5 | 1094 | 9088104 | ||
BC158065.1 | - | 5 | 1094 | 9007364 | ||
BC172029.1 | - | 5 | 1094 | 9054315 |