Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 22, 23 | 3853 | ||
GenBank mRNA | AK128893.1 | - | 2, 3 | 621 | ||
AK129257.1 | - | 16, 17 | 2866 | |||
BC033915.1 | - | 2, 3 | 503 | |||
BC058975.1 | - | 5, 6 | 1567 | |||
BC063268.1 | - | 22, 23 | 3649 | 4187769 | ||
BC080688.1 | - | 22, 23 | 3795 | 6515742 | ||
BC082313.1 | - | 8, 9 | 2008 |