Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048054.2 | NP_001041519.1 | 3 | 973 | ||
NM_130447.3 | NP_569714.2 | 3 | 973 | |||
GenBank mRNA | AB052157.1 | - | 3 | 692 | ||
AF345951.1 | - | 3 | 817 | |||
AF345952.1 | - | 3 | 817 | |||
AF345953.1 | - | 3 | 817 | |||
AF345954.1 | - | 3 | 817 | |||
AK035652.1 | - | 3 | 719 | |||
AK144800.1 | - | 3 | 974 | |||
BC057321.1 | - | 3 | 824 | 6830600 | ||
BC059232.1 | - | 3 | 760 | 6409944 | ||
CN529289.1 | - | 2 | 59 | |||
CV557369.1 | - | 2 | 249 |