Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048054.2 | NP_001041519.1 | 6 | 1506 | ||
NM_130447.3 | NP_569714.2 | 6 | 1506 | |||
GenBank mRNA | AB052157.1 | - | 6 | 1225 | ||
AF345951.1 | - | 6 | 1350 | |||
AF345952.1 | - | 5 | 1185 | |||
AF345953.1 | - | 6 | 1350 | |||
AF345954.1 | - | 5 | 1185 | |||
AK144800.1 | - | 6 | 1507 | |||
BC057321.1 | - | 6 | 1357 | 6830600 | ||
BC059232.1 | - | 6 | 1293 | 6409944 | ||
CF733313.1 | - | 2 | 141 | |||
CN529289.1 | - | 5 | 592 |