Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 4 | 719 | ||
GenBank mRNA | AB055406.1 | - | 4 | 751 | ||
AK018128.1 | - | 4 | 535 | |||
AK036605.1 | - | 4 | 720 | |||
AK042557.1 | - | 4 | 432 | |||
AK047657.1 | - | 4 | 776 | |||
AK086092.1 | - | 4 | 728 | |||
AK138268.1 | - | 4 | 418 | |||
AK160904.1 | - | 4 | 726 | |||
AK161526.1 | - | 4 | 728 | |||
AK162725.1 | - | 4 | 720 | |||
AK163617.1 | - | 4 | 418 | |||
BC026658.1 | - | 4 | 482 | |||
BC116246.1 | - | 4 | 641 | 40093530 | ||
BC116247.1 | - | 4 | 639 | 40093536 |