Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142701.1 | NP_001136173.1 | 20, 21 | 2894 | ||
NM_027521.3 | NP_081797.1 | 19, 20 | 2446 | |||
GenBank mRNA | AK018130.1 | - | 7, 8 | 961 | ||
AK044860.1 | - | 19, 20 | 2408 | |||
AK134029.1 | - | 13, 14 | 1868 | |||
AK151423.1 | - | 20, 21 | 2838 | |||
AK159194.1 | - | 20, 21 | 2895 | |||
AK159474.1 | - | 19, 20 | 2939 | |||
AK161766.1 | - | 12, 13 | 1954 | |||
AK170262.1 | - | 20, 21 | 2880 | |||
AK171304.1 | - | 20, 21 | 2880 | |||
BC039802.1 | - | 1, 2 | 103 | |||
BC053750.1 | - | 19, 20 | 2446 | 6333971 |