Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109985.1 | NP_001103455.1 | 3 | 732 | ||
NM_027528.2 | NP_081804.1 | 3 | 549 | |||
GenBank mRNA | AK018149.1 | - | 3 | 732 | ||
AK045124.1 | - | 3 | 686 | |||
AK053892.1 | - | 3 | 733 | |||
AK138471.1 | - | 3 | 550 | |||
AK162919.1 | - | 3 | 550 |