Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037711.2 | NP_001032800.2 | 2 | 211 | ||
NM_001293727.1 | NP_001280656.1 | 2 | 211 | |||
GenBank mRNA | AK018143.1 | - | 2 | 148 | ||
AK173148.1 | - | 2 | 188 | |||
BC042459.1 | - | 2 | 157 | |||
BC064474.1 | - | 2 | 159 | |||
BC087546.1 | - | 2 | 175 | |||
BC108378.1 | - | 171 | 30736765 |