Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075098.3 | NP_001068566.1 | 6, 7 | 899 | ||
NM_005943.5 | NP_005934.2 | 5, 6 | 903 | |||
NR_033233.1 | 5, 6 | 772 | ||||
XM_011514632.2 | XP_011512934.1 | 6, 7 | 810 | |||
XM_011514633.2 | XP_011512935.1 | 6, 7 | 810 | |||
XM_011514634.2 | XP_011512936.1 | 5, 6 | 707 | |||
XM_017010888.1 | XP_016866377.1 | 5, 6 | 717 | |||
GenBank mRNA | AF034374.1 | - | 6, 7 | 782 | ||
AJ224328.1 | - | 5, 6 | 900 | |||
AK056740.1 | - | 5, 6 | 718 | |||
AK091306.1 | - | 5, 6 | 631 | |||
AK300300.1 | - | 6, 7 | 772 | |||
BC036839.1 | - | 6, 7 | 772 | |||
CX787499.1 | - | 2, 3 | 180 |