Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027541.4 | NP_081817.2 | 6, 7 | 1216 | ||
GenBank mRNA | AK014398.1 | - | 6, 7 | 824 | ||
AK088194.1 | - | 6, 7 | 1217 | |||
AK139852.1 | - | 6, 7 | 849 | |||
AK167435.1 | - | 6, 7 | 853 | |||
BC006782.1 | - | 6, 7 | 801 | 3589226 | ||
BC026607.1 | - | 3, 4 | 970 | |||
BC040787.1 | - | 3, 4 | 970 |