Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 13 | 2174 | ||
NM_001039352.2 | NP_001034441.1 | 13 | 2180 | |||
NM_001039353.2 | NP_001034442.1 | 13 | 2177 | |||
NM_053086.3 | NP_444316.2 | 13 | 2177 | |||
GenBank mRNA | AK014336.1 | - | 13 | 2079 | ||
AK029150.1 | - | 13 | 2025 | |||
AK034817.1 | - | 13 | 2054 | |||
AK129040.1 | - | 13 | 2001 | |||
AK147118.1 | - | 13 | 2050 | |||
AK150332.1 | - | 13 | 2010 | |||
AK151031.1 | - | 13 | 2010 | |||
AK152941.1 | - | 13 | 2011 | |||
AK166753.1 | - | 13 | 2083 |