Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153551.1 | NP_705779.1 | 6 | 420 | ||
GenBank mRNA | AK014482.1 | - | 6 | 425 | ||
AK040537.1 | - | 6 | 421 | |||
AK170891.1 | - | 6 | 418 | |||
BC035208.1 | - | 6 | 420 | 3671384 |