Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081359.3 | NP_001074828.2 | 56 | 8317 | ||
NM_001112721.2 | NP_001106192.1 | 56 | 8299 | |||
GenBank mRNA | AK014485.1 | - | 5 | 1479 | ||
AK122398.1 | - | 33 | 4790 | |||
AY550908.1 | - | 56 | 7907 | |||
BC034075.1 | - | 4 | 2654 | |||
BC047348.1 | - | 47 | 6810 | |||
BC049162.1 | - | 27 | 3997 | |||
BC049224.1 | - | 2 | 107 | |||
BC057458.1 | - | 9 | 1336 | |||
BC057923.1 | - | 8 | 849 | |||
BC064751.1 | - | 6 | 559 |