Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027558.1 | NP_081834.1 | 2, 3 | 600 | ||
GenBank mRNA | AK014543.1 | - | 2, 3 | 499 | ||
AK184727.1 | - | 1, 2 | 150 | |||
AX775753.1 | - | 2, 3 | 556 | |||
BC044759.2 | - | 2, 3 | 541 |