Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 4 | 847 | ||
GenBank mRNA | AK014582.1 | - | 4 | 847 | ||
AK046754.1 | - | 4 | 557 | |||
AK135740.1 | - | 4 | 663 | |||
AK137175.1 | - | 4 | 664 | |||
AK144323.1 | - | 5 | 581 | |||
BC036384.1 | - | 4 | 645 | 1067080 | ||
BC043313.1 | - | 1 | 201 | |||
BC043469.1 | - | 1 | 201 | |||
BC054826.1 | - | 1 | 207 | |||
BC094060.1 | - | 3 | 727 | |||
DQ861995.1 | - | 4 | 511 |