Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 3 | 780 | ||
GenBank mRNA | AK014582.1 | - | 3 | 780 | ||
AK046754.1 | - | 3 | 490 | |||
AK135740.1 | - | 3 | 596 | |||
AK137175.1 | - | 3 | 597 | |||
AK144323.1 | - | 4 | 514 | |||
BC036384.1 | - | 3 | 578 | 1067080 | ||
DQ861995.1 | - | 3 | 444 |