Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921507P07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027564.3 | NP_081840.1 | 3 | 449 | ||
GenBank mRNA | AK014840.1 | - | 3 | 450 | ||
AK161510.1 | - | 3 | 447 | |||
BC055110.1 | - | 3 | 452 | 6705881 |