Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027583.3 | NP_081859.1 | 3, 4 | 895 | ||
NM_029150.4 | NP_083426.1 | 3, 4 | 895 | |||
GenBank mRNA | AK014869.1 | - | 3, 4 | 859 | ||
AK076628.1 | - | 3, 4 | 896 | |||
BC100443.1 | - | 3, 4 | 876 | 30920480 | ||
BC132205.1 | - | 3, 4 | 819 | 40130482 | ||
BC132539.1 | - | 3, 4 | 819 | 40130816 |