Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162979.1 | NP_001156451.1 | 15 | 2225 | ||
GenBank mRNA | AK014876.1 | - | 15 | 2208 | ||
AK029544.1 | - | 16 | 2323 | |||
AV255663.2 | - | 5 | 525 |