Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038616.2 | NP_001033705.2 | 4 | 483 | ||
NM_027591.3 | NP_081867.1 | 6 | 581 | |||
NM_029378.2 | NP_083654.1 | 4 | 395 | |||
GenBank mRNA | AK006548.1 | - | 4 | 498 | ||
AK014934.1 | - | 6 | 582 | |||
BC147175.1 | - | 6 | 553 | 9056223 | ||
BC147176.1 | - | 6 | 553 | 9056216 | ||
DQ316772.1 | - | 6 | 537 | |||
DQ316773.1 | - | 4 | 395 | |||
DQ316774.1 | - | 4 | 395 |