Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142631.1 | NP_001136103.1 | 5 | 491 | ||
NM_029254.1 | NP_083530.1 | 5 | 491 | |||
GenBank mRNA | AK014910.1 | - | 5 | 481 | ||
AK077138.1 | - | 5 | 492 | |||
AY820306.1 | - | 4 | 392 | |||
AY820307.1 | - | 4 | 392 | |||
BC147706.1 | - | 5 | 469 | 9087465 | ||
BC147719.1 | - | 5 | 469 | 9087478 |