Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163507.1 | NP_001156979.1 | 1 | 784 | ||
NM_027613.1 | NP_081889.1 | 1 | 784 | |||
GenBank mRNA | AK014984.1 | - | 1037 | |||
AK036724.1 | - | 1 | 622 | |||
AK137556.1 | - | 1 | 623 | |||
BC137623.1 | - | 1 | 584 | 8860642 | ||
BC144789.1 | - | 1 | 584 | 9053337 |