Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027642.2 | NP_081918.1 | 2 | 329 | ||
GenBank mRNA | AK016480.1 | - | 2 | 330 | ||
AK038320.1 | - | 2 | 249 | |||
AK122560.1 | - | 2 | 259 | |||
AK141804.1 | - | 2 | 272 | |||
AK157045.1 | - | 2 | 330 | |||
AK163345.1 | - | 2 | 285 | |||
AK192342.1 | - | 2 | 167 | |||
AK213676.1 | - | 2 | 167 | |||
BC043127.1 | - | 2 | 277 | 6410361 | ||
BC055330.1 | - | 2 | 272 | 6406483 | ||
BC057374.1 | - | 2 | 235 | 6826091 |