Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033159.2 | NP_001028331.2 | 3 | 1294 | ||
GenBank mRNA | AK016727.1 | - | 3 | 1293 | ||
BC057047.1 | - | 1 | 703 | |||
BC108421.1 | - | 1 | 729 |