Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114311.1 | NP_001107783.1 | 2 | 1615 | ||
NM_001286376.1 | NP_001273305.1 | 2 | 1615 | |||
NM_175162.4 | NP_780371.3 | 2 | 410 | |||
GenBank mRNA | AK030015.1 | - | 2 | 224 | ||
AK053894.1 | - | 2 | 1615 | |||
AK129346.1 | - | 2 | 1575 | |||
AK162847.1 | - | 2 | 444 | |||
BC099949.1 | - | 2 | 424 | 30096824 | ||
BC100572.1 | - | 2 | 410 | 30085660 | ||
BC100573.1 | - | 2 | 410 | 30086796 | ||
BC138237.1 | - | 2 | 181 | 8861256 | ||
BC138238.1 | - | 2 | 181 | 8861257 | ||
BC145211.1 | - | 2 | 181 |