Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000017.3 | NP_000008.1 | 3 | 494 | ||
NM_001302554.1 | NP_001289483.1 | 3 | 494 | |||
GenBank mRNA | AB527081.1 | - | 3 | 346 | ||
AK300645.1 | - | 3 | 372 | |||
BC025963.1 | - | 3 | 393 | 4842286 | ||
M26393.1 | - | 3 | 378 |