Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142691.1 | NP_001136163.1 | 9 | 1327 | ||
GenBank mRNA | AK016891.1 | - | 9 | 1328 | ||
BC116811.1 | - | 8 | 961 | 40126130 | ||
BC116813.1 | - | 8 | 961 | 40126132 |
Gene Symbol: Dmrtc1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142690.1 | NP_001136162.1 | 9 | 1327 |