Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175163.3 | NP_780372.2 | 2 | 563 | ||
GenBank mRNA | AK016830.1 | - | 2 | 399 | ||
AK028417.1 | - | 2 | 526 | |||
AK051667.1 | - | 2 | 377 | |||
BC052084.1 | - | 2 | 563 | 6404620 | ||
BU054587.1 | - | 2 | 563 |