Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028903.2 | NP_083179.2 | 9 | 1721 | ||
GenBank mRNA | AK133176.1 | - | 9 | 1722 | ||
BC016096.1 | - | 9 | 1697 | 4506472 | ||
BC023907.1 | - | 9 | 1637 | 5321978 | ||
BC026758.1 | - | 6 | 747 | |||
BC032159.1 | - | 9 | 1677 | 5354657 | ||
DQ122126.1 | - | 8 | 1371 |