Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027721.2 | NP_081997.1 | 2 | 176 | ||
GenBank mRNA | AK014074.1 | - | 3 | 293 | ||
AK017114.1 | - | 2 | 177 | |||
BC056963.1 | - | 2 | 123 | |||
BC069977.1 | - | 2 | 167 | 30146310 |