Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162955.1 | NP_001156427.1 | 1 | 524 | ||
GenBank mRNA | BC107229.2 | - | 1 | 524 | 40052032 | |
BC107281.2 | - | 1 | 522 | |||
BC107282.2 | - | 1 | 522 | |||
BC119501.1 | - | 1 | 524 | 40052770 | ||
BC119502.1 | - | 1 | 524 | 40052772 | ||
BC119518.1 | - | 1 | 524 | 40055788 |
Set Membership: |
G-protein coupled receptor |