Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077713.1 | NP_001071181.1 | 2 | 335 | ||
GenBank mRNA | AK002229.1 | - | 2 | 317 | ||
AK004417.1 | - | 2 | 265 | |||
AK028031.1 | - | 2 | 306 | |||
AK046436.1 | - | 350 | ||||
BC118927.1 | - | 2 | 335 | 40090186 | ||
BC118928.1 | - | 2 | 342 | 40090189 |