Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 5 | 781 | ||
GenBank mRNA | AF326986.1 | - | 5 | 792 | ||
AK017065.1 | - | 2 | 676 | |||
AK034133.1 | - | 5 | 777 | |||
AK051317.1 | - | 5 | 806 | |||
AK210732.1 | - | 1 | 55 | |||
BC137864.1 | - | 5 | 764 | 8860883 | ||
BC144983.1 | - | 5 | 764 | |||
CV557129.1 | - | 4 | 651 |