Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002438.3 | NP_002429.1 | 23 | 3333 | ||
GenBank mRNA | AK297044.1 | - | 7 | 793 | ||
BC146838.1 | - | 23 | 3256 | 9056765 | ||
DQ663787.1 | - | 23 | 3210 | |||
J05550.1 | - | 23 | 3313 | |||
X55635.1 | - | 23 | 3286 |