Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175164.4 | NP_780373.3 | 8 | 1078 | ||
GenBank mRNA | AK129176.1 | - | 8 | 1016 | ||
BC141394.1 | - | 8 | 947 | 9056007 |