Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111304.1 | NP_001104774.1 | 21 | 4168 | ||
NM_027758.4 | NP_082034.1 | 18 | 3469 | |||
GenBank mRNA | AK017015.1 | - | 7 | 987 | ||
AK049049.1 | - | 18 | 3470 | |||
AK077081.1 | - | 7 | 1001 | |||
AK134614.1 | - | 21 | 3445 | |||
AK173050.1 | - | 21 | 3341 | |||
BC057027.1 | - | 17 | 2756 | 6825869 |